Learn More
Raybiotech Inc Streptavidin Super Mag Magnetic Beads (50 nm) 5ml
Catalog No. NC2888529
Encompass_Preferred
Streptavidin Super Mag (50 nm) are streptavidin conjugated magnetic beads with average particle size of 50 nm. The monolayer of streptavidin that is covalently coupled to their surface makes most of the biotin binding sites sterically available for binding of biotinylated nucleic acids, antibodies, or other biotinylated ligands and targets. With large surface area, excellent colloidal stability and unique surface coating, Streptavidin Super Mag (50 nm) exhibit high binding capacity, low non-specific binding and fast magnetic separation.
Catalog No. NC2888529 Supplier Raybiotech Inc Supplier No. 8SVS505
Streptavidin Super Mag (50 nm) are streptavidin conjugated magnetic beads with average particle size of 50 nm. The monolayer of streptavidin that is covalently coupled to their surface makes most of the biotin binding sites sterically available for binding of biotinylated nucleic acids, antibodies, or other biotinylated ligands and targets. With large surface area, excellent colloidal stability and unique surface coating, Streptavidin Super Mag (50 nm) exhibit high binding capacity, low non-specific binding and fast magnetic separation.