Learn More
Vector Laboratories Streptavidin, DyLight 488 | 1mg
SDP
Supplier:  Vector Laboratories SA54881
Encompass
DyLight 488 Streptavidin can be used to detect biotinylated secondary antibodies and other macromolecules in applications such as immunofluorescence, in situ hybridization, or flow cytometry.
Catalog No. NC1827948