Learn More
Cell Signaling Technology Stress Granule Marker IF Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502727445
Encompass
The Stress Granule Marker IF Antibody Sampler Kit offers an economical solution for evaluating total levels of stress granule-associated proteins. This sampler kit contains a selection of primary antibodies validated for detecting proteins essential for stress granule assembly and function. Each antibody is provided in sufficient quantity to perform multiple western blot experiments.
  • Includes antibodies targeting key stress granule mediators such as G3BP1, G3BP2, TIAR, FMRP, and UBAP2L.
  • Supplied with an HRP-linked secondary antibody for convenient detection.
  • Formulated in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Validated for multiple applications including western blotting, immunofluorescence, immunohistochemistry, and immunoprecipitation.
  • Storage at −20°C is recommended; do not aliquot the antibodies.
Catalog No. 50-272-7445 Supplier Cell Signaling Technology Supplier No. 29530T
The Stress Granule Marker IF Antibody Sampler Kit offers an economical solution for evaluating total levels of stress granule-associated proteins. This sampler kit contains a selection of primary antibodies validated for detecting proteins essential for stress granule assembly and function. Each antibody is provided in sufficient quantity to perform multiple western blot experiments.
  • Includes antibodies targeting key stress granule mediators such as G3BP1, G3BP2, TIAR, FMRP, and UBAP2L.
  • Supplied with an HRP-linked secondary antibody for convenient detection.
  • Formulated in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Validated for multiple applications including western blotting, immunofluorescence, immunohistochemistry, and immunoprecipitation.
  • Storage at −20°C is recommended; do not aliquot the antibodies.