Learn More
Medchemexpress LLC Stressin I TFA | 98.6% | 4472.20 | 5 MG
SDP
Catalog No. 5000424702
Encompass_Preferred
Stressin I TFA (Cyclo(31-34)[DPhe12,Nle21,38,Glu31,Lys34]Ac-hCRF(4-41)) TFA is a potent CRF1 receptor selective agonist with a Ki of 1.7 nM. It induces an increase in adrenocorticotropic hormone (ACTH) levels in rats. This product is for research use only and not sold to patients.
  • Potent CRF1 receptor selective agonist
  • Induces increase in adrenocorticotropic hormone (ACTH) levels
  • White to off-white solid appearance
  • For research use only
Catalog No. 50-004-24702 Supplier Medchemexpress LLC Supplier No. HYP1368A5MG
Stressin I TFA (Cyclo(31-34)[DPhe12,Nle21,38,Glu31,Lys34]Ac-hCRF(4-41)) TFA is a potent CRF1 receptor selective agonist with a Ki of 1.7 nM. It induces an increase in adrenocorticotropic hormone (ACTH) levels in rats. This product is for research use only and not sold to patients.
  • Potent CRF1 receptor selective agonist
  • Induces increase in adrenocorticotropic hormone (ACTH) levels
  • White to off-white solid appearance
  • For research use only