Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Stretch Associates™ Hand Held Stretch Wrap

Catalog No. 501113807
Encompass
Change view
Click to view available options
Width (English) Exterior:
18 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Exterior
50-111-3807 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-111-3807 Supplier MSC Supplier No. 89884746
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Quantity 4 Rolls
Length (English) Exterior 1500 ft.
Width (English) Exterior 18 in.
Applications Dispenser
Color Clear
Type Hand-Held
Description 75 gauge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.