Learn More
Grainger Stretch Wrap Cast Light Duty
Catalog No. 502751697
Encompass_Preferred
The Grainger 18" x 1500' Hand Grade Stretch Wrap Roll is a cast stretch film designed for general purpose pallet wrapping applications. It is constructed from a linear low-density polyethylene and provides good load containment. The film is available in an 18 inch width and 1500 foot length roll. It has 55% pre-stretch capability and is rated for use on light to medium duty loads up to 40 lbs per square inch. The stretch wrap allows for quick, efficient and economical unitizing and palletizing of products.
Catalog No. 50-275-1697 Supplier Grainger Supplier No. 15A983
The Grainger 18" x 1500' Hand Grade Stretch Wrap Roll is a cast stretch film designed for general purpose pallet wrapping applications. It is constructed from a linear low-density polyethylene and provides good load containment. The film is available in an 18 inch width and 1500 foot length roll. It has 55% pre-stretch capability and is rated for use on light to medium duty loads up to 40 lbs per square inch. The stretch wrap allows for quick, efficient and economical unitizing and palletizing of products.