Learn More
Grainger Stretch Wrap Cast Light Duty PK4
Catalog No. 502751759
Encompass_Preferred
The Grainger 15A973 Stretch Wrap is a 120 gauge film made in the USA from linear low density polyethylene, ideal for light to medium duty palletizing and bundling. Available in a 20" x 1500' hand wrap roll with a 2" core, it offers good load containment and stretches up to 200%. The 120 gauge ensures strength and puncture resistance, maximizing load stability and efficiency in warehouses, distribution centers, and manufacturing facilities. Suitable for both automated and hand wrapping systems, it is a cost-effective solution for stabilizing and protecting pallet loads during shipping and storage.
Catalog No. 50-275-1759 Supplier Grainger Supplier No. 15A975
The Grainger 15A973 Stretch Wrap is a 120 gauge film made in the USA from linear low density polyethylene, ideal for light to medium duty palletizing and bundling. Available in a 20" x 1500' hand wrap roll with a 2" core, it offers good load containment and stretches up to 200%. The 120 gauge ensures strength and puncture resistance, maximizing load stability and efficiency in warehouses, distribution centers, and manufacturing facilities. Suitable for both automated and hand wrapping systems, it is a cost-effective solution for stabilizing and protecting pallet loads during shipping and storage.