Learn More
Grainger Stretch Wrap Cast Light Duty PK12
Catalog No. 502751471
Encompass_Preferred
The Grainger 60 Gauge Stretch Wrap is a high-quality film for palletizing and stabilizing loads. Made from linear low-density polyethylene, it offers 270% stretch capability and clings to irregular shapes. Suitable for light to heavy-duty applications, it comes in a 20" x 1500' hand wrap roll. This cost-effective wrap ensures load stability and protection against moisture, dust, and corrosion, making it ideal for securing pallet loads across various industries.
Catalog No. 50-275-1471 Supplier Grainger Supplier No. 15A979
The Grainger 60 Gauge Stretch Wrap is a high-quality film for palletizing and stabilizing loads. Made from linear low-density polyethylene, it offers 270% stretch capability and clings to irregular shapes. Suitable for light to heavy-duty applications, it comes in a 20" x 1500' hand wrap roll. This cost-effective wrap ensures load stability and protection against moisture, dust, and corrosion, making it ideal for securing pallet loads across various industries.