Learn More
Grainger Stretch Wrap Cast Light Duty
Catalog No. 502752178
Encompass_Preferred
The Grainger 60 Gauge Stretch Wrap is a high-quality film for palletizing and stabilizing loads during transport and storage. Made from linear low-density polyethylene, it offers optimal cling, tear resistance, and load stability. Available in various widths and lengths, it features 60 gauge thickness for heavy-duty applications, superior cling for secure wrapping, and excellent optical clarity for load visibility. This industrial-grade stretch wrap ensures reliable performance for pallet load wrapping and protection.
Catalog No. 50-275-2178 Supplier Grainger Supplier No. 15A984
The Grainger 60 Gauge Stretch Wrap is a high-quality film for palletizing and stabilizing loads during transport and storage. Made from linear low-density polyethylene, it offers optimal cling, tear resistance, and load stability. Available in various widths and lengths, it features 60 gauge thickness for heavy-duty applications, superior cling for secure wrapping, and excellent optical clarity for load visibility. This industrial-grade stretch wrap ensures reliable performance for pallet load wrapping and protection.