Learn More
Grainger Stretch Wrap Cast Standard Duty
Catalog No. 502751376
Encompass_Preferred
The Grainger 80 Gauge Stretch Wrap is a high-quality film for palletizing and stabilizing loads during transport and storage. This 15" wide x 1500' long hand wrap offers superior load containment with linear low-density polyethylene, providing optimal cling, tear, and puncture resistance. With an 80-gauge thickness, it stretches up to 200% to secure items on pallets and withstands shipping stresses. Its cling properties allow self-adherence without taping. This cost-effective wrap is ideal for securing pallet loads in warehouses, distribution centers, and shipping departments.
Catalog No. 50-275-1376 Supplier Grainger Supplier No. 15C006
The Grainger 80 Gauge Stretch Wrap is a high-quality film for palletizing and stabilizing loads during transport and storage. This 15" wide x 1500' long hand wrap offers superior load containment with linear low-density polyethylene, providing optimal cling, tear, and puncture resistance. With an 80-gauge thickness, it stretches up to 200% to secure items on pallets and withstands shipping stresses. Its cling properties allow self-adherence without taping. This cost-effective wrap is ideal for securing pallet loads in warehouses, distribution centers, and shipping departments.