Learn More
Grainger Stretch Wrap Cast Standard Duty
Catalog No. 502751693
Encompass_Preferred
The Grainger 15C003 Stretch Wrap is a 90 gauge stretch film that provides load containment and product protection. It is constructed from a linear low-density polyethylene and is available in a 20" x 1500' hand wrap roll. Key features include 90 gauge thickness for medium duty applications, linear low-density polyethylene material for strength and cling, and machine or hand wrap versatility. The stretch wrap is designed to secure pallet loads up to 2000 lbs during shipping and storage. It conforms and adheres to irregular shapes and is perforated every 18" for easy hand wrapping. This industrial stretch wrap offers a cost-effective solution for unitizing and protecting pallet loads.
Catalog No. 50-275-1693 Supplier Grainger Supplier No. 15C003
The Grainger 15C003 Stretch Wrap is a 90 gauge stretch film that provides load containment and product protection. It is constructed from a linear low-density polyethylene and is available in a 20" x 1500' hand wrap roll. Key features include 90 gauge thickness for medium duty applications, linear low-density polyethylene material for strength and cling, and machine or hand wrap versatility. The stretch wrap is designed to secure pallet loads up to 2000 lbs during shipping and storage. It conforms and adheres to irregular shapes and is perforated every 18" for easy hand wrapping. This industrial stretch wrap offers a cost-effective solution for unitizing and protecting pallet loads.