Learn More
Grainger Stretch Wrap Cast Standard Duty
Catalog No. 502752142
Encompass_Preferred
The Grainger 80 Gauge Stretch Wrap is a high-quality stretch film designed for palletizing and stabilizing loads during transport. This 15" wide film features superior load containment and is constructed with a linear low-density polyethylene material that provides optimal cling and wrap force. With a thickness of 80 gauge (0.0008"), it is durable yet lightweight. The stretch wrap is available in convenient 30" x 1500' hand wrap rolls and can stretch up to 200% to tightly secure items on pallets. It is an ideal solution for stabilizing and protecting pallet loads during shipping and storage.
Catalog No. 50-275-2142 Supplier Grainger Supplier No. 15C004
The Grainger 80 Gauge Stretch Wrap is a high-quality stretch film designed for palletizing and stabilizing loads during transport. This 15" wide film features superior load containment and is constructed with a linear low-density polyethylene material that provides optimal cling and wrap force. With a thickness of 80 gauge (0.0008"), it is durable yet lightweight. The stretch wrap is available in convenient 30" x 1500' hand wrap rolls and can stretch up to 200% to tightly secure items on pallets. It is an ideal solution for stabilizing and protecting pallet loads during shipping and storage.