Learn More
Grainger Stretch Wrap Cast Standard Duty
Catalog No. 502753339
Encompass_Preferred
The Goodwrappers Stretch Wrap 70 ga Gauge is a 70 gauge stretch film that provides load containment and product protection. It is constructed from a linear low-density polyethylene and is available in hand and machine grades. This stretch wrap film comes in a variety of widths and lengths and can be purchased by the roll. It is designed to cling to itself and not the load. The 70 gauge offers good puncture resistance and is recommended for light to medium duty applications. This product helps secure pallet loads for transport and storage.
Catalog No. 50-275-3339 Supplier Grainger Supplier No. 15A985
The Goodwrappers Stretch Wrap 70 ga Gauge is a 70 gauge stretch film that provides load containment and product protection. It is constructed from a linear low-density polyethylene and is available in hand and machine grades. This stretch wrap film comes in a variety of widths and lengths and can be purchased by the roll. It is designed to cling to itself and not the load. The 70 gauge offers good puncture resistance and is recommended for light to medium duty applications. This product helps secure pallet loads for transport and storage.