Learn More
Grainger Stretch Wrap Cast Standard Duty PK12
Catalog No. 502750509
Encompass_Preferred
The Grainger 18" x 1500' Hand Grade Stretch Wrap Roll is a cast stretch film designed for general purpose pallet wrapping applications. It is constructed from polyethylene and provides good load containment. The 18" width allows it to be used manually without a dispenser. It can wrap up to 50 pallets when used with a dispenser. The film is available in a 1500' length roll which provides extended use between roll changes. The cast film is designed to cling to itself and contains no tackifiers. This general purpose pallet wrap provides economical load stabilization.
Catalog No. 50-275-0509 Supplier Grainger Supplier No. 15A981
The Grainger 18" x 1500' Hand Grade Stretch Wrap Roll is a cast stretch film designed for general purpose pallet wrapping applications. It is constructed from polyethylene and provides good load containment. The 18" width allows it to be used manually without a dispenser. It can wrap up to 50 pallets when used with a dispenser. The film is available in a 1500' length roll which provides extended use between roll changes. The cast film is designed to cling to itself and contains no tackifiers. This general purpose pallet wrap provides economical load stabilization.