Learn More
IBA LifeSciences StrGate Acptr Vctr pASG-IBA44
Supplier:  IBA LifeSciences 54044001
Encompass_Preferred
The pASG-IBA44 vector is designed for expression of recombinant proteins with an N-terminal Strep-tagII and a C-terminal His-tag in E. coli. The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, the inducible tetracycline promoter/operator for the regulated expression of proteins, and the ompA signal for periplasmic secretion of the recombinant protein. Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity. During the secretion into the periplasmic space the ompA signal sequence will be cleaved off.
Catalog No. NC3566358