Learn More
IBA LifeSciences StrGate Acptr Vctr pCSG-IBAwt2
Supplier:  IBA LifeSciences 55001001
Encompass_Preferred
pCSG-IBAwt2 is a large expression vector with universal features for transient expression as well as for generation of stable mammalian cell lines. The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells and ColE1 origin for a high plasmid copy number. Extrachromosomal replication in mammalian cells could occur either by origin of replication from Epstein-Barr Virus (oriP) or by SV40 ori. For the former the vector provides the EBNA-1 gene and for the latter the cell line has to be latently infected with SV40 or express the SV40 large T antigen (e.g. HEK293T, COS-1, COS-7). Stable cell lines can be selected by the neomycin resistance gene (NeoR). In addition, the human cytomegalovirus (CMV) immediate-early promoter enables a high-level expression. During translocation from the cytosol the signal peptide is removed from the protein by endogenous proteases.
Catalog No. NC3566450