Learn More
MTC Bio Storage Box, hinged lid, 100 x 1.5ml, Black
Supplier:  MTC Bio R1020BK
Encompass_Preferred
This item has a minimum qty of 4 per supplier requirements.
Test Tube Racks MTC Bios benchtop tube racks are made of 100% polypropylene and are available in three popular configurations. No. R1030 is a 4-way rack that can be used to hold 4x 50ml, 12x 15ml or 32x micro tubes (0.5, 1.5 or 2ml). They can be snap-linked together to form as large as rack as is needed. No. R1070 is a unique rack that holds 30x 15ml and 20x 50ml. No R1050 is a reversible rack for 96x 1.5ml on one side and 96x 0.5ml on the other. Freezer/Storage Boxes Suitable for refrigerator and freezer storage as low as -80C, these boxes have translucent lids and numbered cavities. No. R1010 is a PCR workstation rack/box for 96x 0.2ml (plates, strips or individual tubes). No. R1020 is a 1010 format with an attached hinged lid. No. R1060 is the standard 99 format with a friction fit lid.
Catalog No. 50-112-7254