Learn More
Chemglass Life Sciences 100mL Storage Vessel, Cajon, Airfree, Schlenk, 0-8mm Valve, 235mm OAH
SDP
Catalog No. AF009603
Encompass_Preferred
100mL Storage Vessel, Cajon, Airfree, Schlenk, 0-8mm Valve, 235mm OAH. Storage vessel sidearm has a 9mm OD tubulation for use with 3/8" Cajon connections. Chem-Vac™ Chem-Cap valve is used to control gas or liquid flow into the vessel.
Catalog No. AF009603 Supplier Chemglass Life Sciences Supplier No. AF009603
100mL Storage Vessel, Cajon, Airfree, Schlenk, 0-8mm Valve, 235mm OAH. Storage vessel sidearm has a 9mm OD tubulation for use with 3/8" Cajon connections. Chem-Vac™ Chem-Cap valve is used to control gas or liquid flow into the vessel.