Learn More
MSC Diversey Neutral Cleaner, Citrus, 1 gal, Green Seal Certified, 4/Carton
Catalog No. 502817726
Encompass
Diversey Stride Neutral Cleaner, Citrus Scent, is a concentrated formula designed for the everyday cleaning of floors and other hard surfaces. Its non-alkaline composition prevents dulling of floor finishes. This versatile cleaner can be utilized with automatic scrubbers, mop buckets, and spray bottles, and its low-foaming formula minimizes recovery tank emptying.
  • Concentrated formula for daily cleaning
  • Non-alkaline formula does not dull floor finishes
  • Versatile for use in automatic scrubbers, mop buckets, and spray bottles
  • No rinsing required
  • Low-foaming formula for increased productivity and less recovery tank emptying
  • Cleans floors and other hard surfaces
Catalog No. 50-281-7726 Supplier MSC Supplier No. 03721495
Diversey Stride Neutral Cleaner, Citrus Scent, is a concentrated formula designed for the everyday cleaning of floors and other hard surfaces. Its non-alkaline composition prevents dulling of floor finishes. This versatile cleaner can be utilized with automatic scrubbers, mop buckets, and spray bottles, and its low-foaming formula minimizes recovery tank emptying.
  • Concentrated formula for daily cleaning
  • Non-alkaline formula does not dull floor finishes
  • Versatile for use in automatic scrubbers, mop buckets, and spray bottles
  • No rinsing required
  • Low-foaming formula for increased productivity and less recovery tank emptying
  • Cleans floors and other hard surfaces