Learn More
MSC Diversey Neutral Cleaner, Concentrated, pH 7, 5 gal., 1 Box
Catalog No. 502817498
Encompass
This neutral cleaner is a water-based, concentrated solution designed for floor cleaning, stripping, and sealing. It is suitable for various materials including vinyl, laminate, linoleum, and wood, and comes in a 5.00-gallon box with a neutral pH of 7.
  • Citrus scent
  • Low-foaming formula allows for less emptying of recovery tanks
Catalog No. 50-281-7498 Supplier MSC Supplier No. 03721560
This neutral cleaner is a water-based, concentrated solution designed for floor cleaning, stripping, and sealing. It is suitable for various materials including vinyl, laminate, linoleum, and wood, and comes in a 5.00-gallon box with a neutral pH of 7.
  • Citrus scent
  • Low-foaming formula allows for less emptying of recovery tanks