Learn More
Cayman Chemical STRIfIed STRyl GlucosIdS 10mg
SDP
Catalog No. 502938854
Encompass_Preferred
A mixture of esterified steryl glucosides; found in vegetables, vegetable oils, fruits, cereals, legumes, fungi, and algae and as bioactive lipids in pre-germinated brown rice; contains a mixture of β-sitosteryl glucoside, β-campesteryl glucoside, and β-stigmasteryl glucoside
Catalog No. 50-293-8854 Supplier Cayman Chemical Supplier No. 2720410 MG
A mixture of esterified steryl glucosides; found in vegetables, vegetable oils, fruits, cereals, legumes, fungi, and algae and as bioactive lipids in pre-germinated brown rice; contains a mixture of β-sitosteryl glucoside, β-campesteryl glucoside, and β-stigmasteryl glucoside