Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Striker Bolt For FastPrep-24™

Catalog No. MP11600508 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP11600508 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP11600508 Supplier MP Biomedicals™ Supplier No. 116005508
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Striker Bolt is a part for the FastPrep-24™ 5G.

  • Samples and buffers are added to a 2.0 mL Lysing Matrix tube containing specialized Lysing Matrix beads.
  • The ergonomic design of the instrument ensures an easy loading of the sample tubes that remains securely sealed during the processing.
  • The homogenization speed and duration times are manually programmed or accessed from onboard memory.
  • After setting your speed and time with the touch of a button, just push run, and in less than a minute your samples are completely lysed.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) FastPrep-24 5G
Quantity 1 Ea.

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.