Learn More
Alkali Scientific Classic™ Sterile, Racked Pipet Tips, Universal Fit, 1000µl, Racks of 96, 960/pk, 4800/cs
SDP
Catalog No. 502335820
Encompass_Preferred
Classic™ Sterile Racked Pipette Tips are deisgned to fit a wide variety of research grade pipettors. Tips are standard 9mm "on center" to ensure a perfect fit. Semi-flexible walls and a series of subtle sealing rings help ensure a secure fit to improve overall pipetting accuracy. Seating tips and ejecting tips becomes an easier task with Alkali Scientific sterile racked pipette tips. They are certified Rnase, Dnase and pyrogen free and withstand organic solvents.
Catalog No. 50-233-5820 Supplier Alkali Scientific Supplier No. ST3000CS
Classic™ Sterile Racked Pipette Tips are deisgned to fit a wide variety of research grade pipettors. Tips are standard 9mm "on center" to ensure a perfect fit. Semi-flexible walls and a series of subtle sealing rings help ensure a secure fit to improve overall pipetting accuracy. Seating tips and ejecting tips becomes an easier task with Alkali Scientific sterile racked pipette tips. They are certified Rnase, Dnase and pyrogen free and withstand organic solvents.