Learn More
Med Vet International Miltex instrument Stringer, 2.5in X 6in, With Side Lock, 3-75
Supplier:  Med Vet International MIL375
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. This is a Miltex Instrument Stringer. Stringers are designed to keep hinged instruments in an open position during sterilization. Stringers help keep instruments organized and this helps you maximize usable tray/container space. Specifications: 2.5in X 6in, with Side Lock
Catalog No. 50-264-3540