Learn More
Electron Microscopy Sciences EMS Strip Aperture, 20-30-30-50 µm holes, 31.0 x 5.0 x 0.10 mm
SDP
Catalog No. 50980840
Encompass_Preferred
Strip apertures made from Platinum-Iridium (95:5).
Holes: 20-30-30-50 µm
Dimensions: 31.0 x 5.0 x 0.10 mm
Catalog No. 50-980-840 Supplier Electron Microscopy Sciences Supplier No. 4921401
Strip apertures made from Platinum-Iridium (95:5).
Holes: 20-30-30-50 µm
Dimensions: 31.0 x 5.0 x 0.10 mm