Learn More
Electron Microscopy Sciences EMS Strip Aperture, 20-60-100 µm holes, 10.0 x 2.5 x 0.10 mm
SDP
Catalog No. 50980839
Encompass_Preferred
Strip apertures made from Platinum-Iridium (95:5).
Holes: 20-60-100 µm
Dimensions: 10.0 x 2.5 x 0.10 mm
Catalog No. 50-980-839 Supplier Electron Microscopy Sciences Supplier No. 4921301
Strip apertures made from Platinum-Iridium (95:5).
Holes: 20-60-100 µm
Dimensions: 10.0 x 2.5 x 0.10 mm