Learn More
Electron Microscopy Sciences EMS Strip Aperture Hitachi, 10-10-20-70 µm holes, 41.0 x 4.0 x 0.10 mm
SDP
Catalog No. 50980831
Encompass_Preferred
Strip apertures made from Platinum-Iridium (95:5).
Holes: 10-10-20-70 µm
Dimensions: 41.0 x 4.0 x 0.10 mm
Catalog No. 50-980-831 Supplier Electron Microscopy Sciences Supplier No. 4921101
Strip apertures made from Platinum-Iridium (95:5).
Holes: 10-10-20-70 µm
Dimensions: 41.0 x 4.0 x 0.10 mm