Learn More
Electron Microscopy Sciences EMS Strip Aperture Hitachi, 10-30-50-70 µm holes, 45.0 x 4.0 x 0.10 mm
SDP
Catalog No. 50980838
Encompass_Preferred
Strip apertures made from Platinum-Iridium (95:5).
Holes: 10-30-50-70 µm
Dimensions: 45.0 x 4.0 x 0.10 mm
Catalog No. 50-980-838 Supplier Electron Microscopy Sciences Supplier No. 4921203
Strip apertures made from Platinum-Iridium (95:5).
Holes: 10-30-50-70 µm
Dimensions: 45.0 x 4.0 x 0.10 mm