Learn More
Electron Microscopy Sciences EMS Strip Aperture Hitachi, 30-50-70-100 µm holes, 41.0 x 4.0 x 0.10 mm
SDP
Catalog No. 50980834
Encompass_Preferred
Strip apertures made from Platinum-Iridium (95:5).
Holes: 30-50-70-100 µm
Dimensions: 41.0 x 4.0 x 0.10 mm
Catalog No. 50-980-834 Supplier Electron Microscopy Sciences Supplier No. 4921104
Strip apertures made from Platinum-Iridium (95:5).
Holes: 30-50-70-100 µm
Dimensions: 41.0 x 4.0 x 0.10 mm