Learn More
Electron Microscopy Sciences EMS Strip Aperture JEOL, 30-50-150 µm holes, 9.0 x 2.35 x 0.10 mm
SDP
Catalog No. 50980829
Encompass_Preferred
Strip apertures made from Platinum-Iridium (95:5).
Holes: 30-50-150 µm
Dimensions: 9.0 x 2.35 x 0.10 mm
Catalog No. 50-980-829 Supplier Electron Microscopy Sciences Supplier No. 4921003
Strip apertures made from Platinum-Iridium (95:5).
Holes: 30-50-150 µm
Dimensions: 9.0 x 2.35 x 0.10 mm