Learn More
Med Vet International Curad Medi-Strip Reinforced Wound Closures, 1/4in X 3in, 3/Pack
Catalog No. 502831230
Encompass_Preferred
Adhesive wound closures with an exceptionally porous spun-bond fiber backing, includes bilateral nylon filaments for high tensile strength. Medi-Strips are used in addition to, or in place of, sutures Feature exceptionally porous spunbound fiber backing with reinforced nylon filaments for strength May stay in place for up to 7 days The hypoallergenic adhesive allows for excellent adhesion while still being gentle on skin Each size is supplied in color-coded packaging for quick identification1/4in X 3in3 per pack Comparable to products like ZipStitch instant stitches Available without a prescription Non-Invasive Easy to use Less pain than Stitches and Staples Less Scarring than Stitches Faster than Stitches Avoid cost of Stitches or Staples Rapid and effective wound closure Small and easy to transport Great addition to first aid kits
Catalog No. 50-283-1230 Supplier Med Vet International Supplier No. SS314
Adhesive wound closures with an exceptionally porous spun-bond fiber backing, includes bilateral nylon filaments for high tensile strength. Medi-Strips are used in addition to, or in place of, sutures Feature exceptionally porous spunbound fiber backing with reinforced nylon filaments for strength May stay in place for up to 7 days The hypoallergenic adhesive allows for excellent adhesion while still being gentle on skin Each size is supplied in color-coded packaging for quick identification1/4in X 3in3 per pack Comparable to products like ZipStitch instant stitches Available without a prescription Non-Invasive Easy to use Less pain than Stitches and Staples Less Scarring than Stitches Faster than Stitches Avoid cost of Stitches or Staples Rapid and effective wound closure Small and easy to transport Great addition to first aid kits