Learn More
Med Vet International 3M Attest Sterilization Chemical Integrator Strip, Steam, 2 inch, 1243A, 2/Case
Catalog No. 502829688
Encompass_Preferred
2-inch Comply SteriGage Chemical Integrator identifies whether the conditions for sterilization are being met during steam sterilization Easy-to-read display window shows whether sterilization is a success or failure SteriGage Steam Chemical Integrator has a chemical pellet that offers visual indication along the color bar It maintains consistent performance across all 250 – 275° F steam sterilization cycles An extender strip helps with the retrieval of processed indicators
Catalog No. 50-282-9688 Supplier Med Vet International Supplier No. 134499CS
2-inch Comply SteriGage Chemical Integrator identifies whether the conditions for sterilization are being met during steam sterilization Easy-to-read display window shows whether sterilization is a success or failure SteriGage Steam Chemical Integrator has a chemical pellet that offers visual indication along the color bar It maintains consistent performance across all 250 – 275° F steam sterilization cycles An extender strip helps with the retrieval of processed indicators