Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Stripettor™ Pro Pipet Controller
GREENER_CHOICE

Catalog No. 07000512 Shop All Corning Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-512 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-512 Supplier Corning™ Supplier No. 4900
  • PRODUCT PROMOTION
    $467.00
    $333.00 / Case of 1

    Save $134.00

    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pipet controller with LCD display for continuous visualization of speed, modes, and battery status.

This light, ergonomic, motorized pipet controller is designed for use with glass and plastic serological pipets from 0.5 to 100 mL. Conveniently positioned switches change operating modes and speeds quickly to handle different liquid volumes and various viscosities. Aspirating and dispensing speed is controlled through the finger triggers. When fully charged, the batteries enable up to 6 hours of continuous use. The large LCD display clearly indicates battery status, pipetting mode, and speed.

  • LCD display for continuous visualization of speed, modes, and battery status
  • Multiple speed options for both aspiration and dispensing
  • Well positioned function buttons
  • Lithium ion battery
  • 3-position charging stand (4999 only)

Corning Stripettor Pro Pipet Controller comes with a universal power supply, 0.2 μm filter, and a wall hanger.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Color White
Disposable Reusable
For Use With Serological Pipettes
Number of Channels 1
Sterility Non-sterile
Type Pipette Controller
Volume (Metric) 0.5 to 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.