Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Stripettor™ Ultra Charging Stand
GREENER_CHOICE

Catalog No. 07201127 Shop All Corning Products
Change view
Click to view available options
9 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
07-201-127 9 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-201-127 Supplier Corning™ Supplier No. 4998
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Corning™ Stripettor™ Ultra Charging Stand is intended to conveniently charge and store the pipette controller when not in use. There are three possible positions for either storing or charging, all are visible as icons printed on the stand.

  • Break position: Convenient while working, helps to avoid contamination
  • Rest position: Ideal way to store a pipette controller after use
  • Charging position: Use while pipet controller is in charging process
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Marked
Description Charging Stand
Autoclavable Not Autoclavable
Color Gray
Disposable Reusable
Material Polypropylene
Sterility Non-sterile
For Use With (Equipment) Corning Stripettor Ultra Pipet Controller (4099)
Voltage 9 V
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.