Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Stripettor™ Ultra Pipet Controller
SCI_ED

Catalog No. S72302 Shop All Corning Products
Change view
Click to view available options
Pipet Controller
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S72302 Pipet Controller
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S72302 Supplier Corning™ Supplier No. 4099/EMD
This item is not returnable. View return policy

A light, ergonomic, motorized pipet controller designed for use with glass and plastic serological pipets from 0.5 to 100mL

Conveniently positioned switches change operating modes and speeds quickly to handle different liquid volumes and various viscosities. Aspirating and dispensing speed is controlled through the finger triggers. When fully charged, the batteries enable up to 8 hours of continuous use.
  • LCD display for continuous visualization of speed, modes, and battery status
  • Additional colored nose cones to customize by application or user
  • Quiet powerful motor
  • Well positioned function buttons
TRUSTED_SUSTAINABILITY

Specifications

Battery Type AAA
Type Pipette Controller
Display LCD
Filter Type 0.2μm
For Use With (Equipment) Glass and plastic serological pipets from 0.5 to 100 mL
Includes Universal power supply, three 0.2μm filters, three colored pieces and wall hanger
Number of Speeds 2
Quantity 1
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.