Learn More
Biotang Inc Striping Buffer3 (4X) Glycine 0.4M SDS 0.2% Tween-20 2% pH 2.5, 250 ml
Catalog No. 50152189 Shop All BIOTANG Inc Products
Encompass_Preferred
Striping Buffer3 (4X) Glycine 0.4M SDS 0.2% Tween-20 2% pH 2.5, 250 ml.
Catalog No. 50-152-189 Supplier Biotang Inc Supplier No. BTBB716
Striping Buffer3 (4X) Glycine 0.4M SDS 0.2% Tween-20 2% pH 2.5, 250 ml.