Learn More
Bioworld Blot Stripping Buffer II (4X), 250 mL
Catalog No. 50198548
Encompass_Preferred
Product DescriptionBlot stripping buffer II (4x) contains enough liquid concentrate to make 4L of 1X solution. Product is 0.2M glycine and has a pH value of 2.5.To use this product, place the membrane into stripping buffer for 30-45 minutes with gentle agitation. Buffer should be heated to approximately 50°C for incubation. Wash membrane 3 times with TBST or PBST before reprobing.Product is ultra pure grade.
Catalog No. 50-198-548 Supplier Bioworld Supplier No. 209600051
Product DescriptionBlot stripping buffer II (4x) contains enough liquid concentrate to make 4L of 1X solution. Product is 0.2M glycine and has a pH value of 2.5.To use this product, place the membrane into stripping buffer for 30-45 minutes with gentle agitation. Buffer should be heated to approximately 50°C for incubation. Wash membrane 3 times with TBST or PBST before reprobing.Product is ultra pure grade.