Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Stripwell™ Microplates
GREENER_CHOICE

Catalog No. 07200315 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Clear
White
Binding Property:
High Binding
Total Volume (Metric) Well:
190 μL
360 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Binding Property Total Volume (Metric) Well
07-200-315 White High Binding 190 μL
07-200-596 Black High Binding 360 μL
07-200-291 Clear High Binding 190 μL
07-200-31 Clear High Binding 360 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07-200-315 Supplier Corning™ Supplier No. 2485
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.

Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.

  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.

Order Info

Bulk packed. Cat. No. 07-200-24 has individual 1 x 8 strips without frame.

Certifications

Cat. Nos. 07-200-31 and -32 have certified surface chemistry.

TRUSTED_SUSTAINABILITY

Specifications

Binding Property High Binding
Format Strip Plate
Lid Without Lid
Array 8 x 12
Packaging 4 inner packs of 25
Binding Type High Bind
Color White
Product Type Microplate Strip
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 125 μL
Sterility Non-sterile
Surface Treatment High Binding
Total Volume (Metric) Well 190 μL
Well Shape Round
For Use With (Application) In-vitro diagnostic assays
No. per Case 100/Cs.
No. per Pack 25/Pk.
Quantity 100/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.