Learn More
Benchmark Products Inc Benchmark Products B-3648-1S Sterile Trash Bags, 36 in x 48 in, Validated Sterile, 250 Bags/Case
SDP
Catalog No. NC1207195
Encompass
The BENCHMARK PRODUCTS INC sterile trash bag is specifically designed for critical cleanroom environments, measuring 36 in x 48 in with a robust 1 mil thickness. Manufactured in an ISO Class 5 cleanroom, it ensures a validated sterility level of 10⁻⁶, making it ideal for scientific and pharmaceutical waste containment. The bags come double-bagged to facilitate safe entry into sterile areas, differentiating them from standard trash bags and ensuring maximum safety.
Catalog No. NC1207195 Supplier Benchmark Products Inc Supplier No. B36481S
The BENCHMARK PRODUCTS INC sterile trash bag is specifically designed for critical cleanroom environments, measuring 36 in x 48 in with a robust 1 mil thickness. Manufactured in an ISO Class 5 cleanroom, it ensures a validated sterility level of 10⁻⁶, making it ideal for scientific and pharmaceutical waste containment. The bags come double-bagged to facilitate safe entry into sterile areas, differentiating them from standard trash bags and ensuring maximum safety.