Learn More
Electron Microscopy Sciences Sterile Petri Dish 35x10 mm, 9 ml, 35x10 mm
SDP
Catalog No. 501900273
Encompass_Preferred
Our Petri Dishes are available in a variety of shapes and sizes for use in routine procedures and with automated equipment. Economical, optical clarity, dishes are precision-molded from biomedical grade polystyrene so cultures are clearly visible without distortion. Dishes are packaged 20 each in sturdy polyethylene sleeves. These dishes are not recommended for tissue culture application, due to non treated by "corona effect". Made of polystyrene. For unimpaired observation of specimen growth. Three venting ribs to prevent condensation built-up.
Volume: 9 ml
Size: 35 x 10 mm
Sterile: Sterile
Catalog No. 50-190-0273 Supplier Electron Microscopy Sciences Supplier No. 7067870
Our Petri Dishes are available in a variety of shapes and sizes for use in routine procedures and with automated equipment. Economical, optical clarity, dishes are precision-molded from biomedical grade polystyrene so cultures are clearly visible without distortion. Dishes are packaged 20 each in sturdy polyethylene sleeves. These dishes are not recommended for tissue culture application, due to non treated by "corona effect". Made of polystyrene. For unimpaired observation of specimen growth. Three venting ribs to prevent condensation built-up.
Volume: 9 ml
Size: 35 x 10 mm
Sterile: Sterile