Learn More
Electron Microscopy Sciences Sterile Petri Dish 60x9 mm, 12 ml, 50x8 mm
SDP
Catalog No. 501900272
Encompass_Preferred
Our Petri Dishes are available in a variety of shapes and sizes for use in routine procedures and with automated equipment. Economical, optical clarity, dishes are precision-molded from biomedical grade polystyrene so cultures are clearly visible without distortion. Dishes are packaged 20 each in sturdy polyethylene sleeves. These dishes are not recommended for tissue culture application, due to non treated by "corona effect". Made of polystyrene. Box type dish with tight lid ideal for culturing of mycobacteria aerosol testing, membrane filter and immunodiffusion.
Volume: 12 ml
Size: 50 x 8 mm
Sterile: Sterile
Catalog No. 50-190-0272 Supplier Electron Microscopy Sciences Supplier No. 7067860
Our Petri Dishes are available in a variety of shapes and sizes for use in routine procedures and with automated equipment. Economical, optical clarity, dishes are precision-molded from biomedical grade polystyrene so cultures are clearly visible without distortion. Dishes are packaged 20 each in sturdy polyethylene sleeves. These dishes are not recommended for tissue culture application, due to non treated by "corona effect". Made of polystyrene. Box type dish with tight lid ideal for culturing of mycobacteria aerosol testing, membrane filter and immunodiffusion.
Volume: 12 ml
Size: 50 x 8 mm
Sterile: Sterile