Learn More
Electron Microscopy Sciences Sterile Petri Dish 60x15 mm, 28 ml, 55x13 mm
SDP
Catalog No. 501900271
Encompass_Preferred
Our Petri Dishes are available in a variety of shapes and sizes for use in routine procedures and with automated equipment. Economical, optical clarity, dishes are precision-molded from biomedical grade polystyrene so cultures are clearly visible without distortion. Dishes are packaged 20 each in sturdy polyethylene sleeves. These dishes are not recommended for tissue culture application, due to non treated by "corona effect". Made of polystyrene. For unimpaired observation of specimen growth. Three venting ribs help prevent condensation built-up.
Volume: 28 ml
Size: 55 x 13 mm
Sterile: Sterile
Catalog No. 50-190-0271 Supplier Electron Microscopy Sciences Supplier No. 7067850
Our Petri Dishes are available in a variety of shapes and sizes for use in routine procedures and with automated equipment. Economical, optical clarity, dishes are precision-molded from biomedical grade polystyrene so cultures are clearly visible without distortion. Dishes are packaged 20 each in sturdy polyethylene sleeves. These dishes are not recommended for tissue culture application, due to non treated by "corona effect". Made of polystyrene. For unimpaired observation of specimen growth. Three venting ribs help prevent condensation built-up.
Volume: 28 ml
Size: 55 x 13 mm
Sterile: Sterile