Learn More
Electron Microscopy Sciences Sterile Petri Dish 100x25 mm, 125 ml, 90x25 mm
SDP
Catalog No. 501900270
Encompass_Preferred
Our Petri Dishes are available in a variety of shapes and sizes for use in routine procedures and with automated equipment. Economical, optical clarity, dishes are precision-molded from biomedical grade polystyrene so cultures are clearly visible without distortion. Dishes are packaged 20 each in sturdy polyethylene sleeves. These dishes are not recommended for tissue culture application, due to non treated by "corona effect". Made of polystyrene. Similar to D210-8 but with no-vented lid having a raised straight ridge around top for stable stacking.
Volume: 125 ml
Size: 90 x 25 mm
Sterile: Sterile
Catalog No. 50-190-0270 Supplier Electron Microscopy Sciences Supplier No. 7067840
Our Petri Dishes are available in a variety of shapes and sizes for use in routine procedures and with automated equipment. Economical, optical clarity, dishes are precision-molded from biomedical grade polystyrene so cultures are clearly visible without distortion. Dishes are packaged 20 each in sturdy polyethylene sleeves. These dishes are not recommended for tissue culture application, due to non treated by "corona effect". Made of polystyrene. Similar to D210-8 but with no-vented lid having a raised straight ridge around top for stable stacking.
Volume: 125 ml
Size: 90 x 25 mm
Sterile: Sterile