Learn More
Cole-Parmer Argos Technologies Sterile Sampling Scoops, 4 oz (120 mL); 100/Cs
Catalog No. 501960573 Shop All Cole-Parmer Products
Encompass_Preferred
These sampling scoops are manufactured from sturdy one-piece polystyrene plastic which can be used to hold a variety of media. These disposable scoops were designed with an ergonomic, comfortably curved handle, that makes sample retrieval easy. The flat bottom surface helps prevent spillage and allows the scoop to be used as a weighing dish.
Catalog No. 50-196-0573 Supplier Cole-Parmer Supplier No. 0626002
These sampling scoops are manufactured from sturdy one-piece polystyrene plastic which can be used to hold a variety of media. These disposable scoops were designed with an ergonomic, comfortably curved handle, that makes sample retrieval easy. The flat bottom surface helps prevent spillage and allows the scoop to be used as a weighing dish.