Learn More
Med Vet International Dextrolyte Sterile Solution Injection, Electrolytes And Dextrose, 1000mL, Each
Catalog No. 502827812
Encompass_Preferred
This sterile solution of electrolytes and dextrose is for use in conditions associated with fluid and electrolyte loss, such as dehydration, shock, vomiting and diarrhea, particularly when an immediate source of energy is also indicated. For use in adult cattle and horses, calves, ponies, foals, adult sheep and swine. Administer by intravenous or intraperitoneal injection; do not administer intraperitoneally to horses.
Catalog No. 50-282-7812 Supplier Med Vet International Supplier No. RXDEXT1000INJV
This sterile solution of electrolytes and dextrose is for use in conditions associated with fluid and electrolyte loss, such as dehydration, shock, vomiting and diarrhea, particularly when an immediate source of energy is also indicated. For use in adult cattle and horses, calves, ponies, foals, adult sheep and swine. Administer by intravenous or intraperitoneal injection; do not administer intraperitoneally to horses.