Learn More
Med Vet International Sterile Water For Injection, Diluent, 1000mL, Each
Catalog No. 502828425
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Sterile water for injection is suitable for use as a diluent for the preparation of pharmaceutical solutions. Single-dose vial contains no preservatives. Not suitable for IV injection without first having been made isotonic by addition of suitable solutes.
Catalog No. 50-282-8425 Supplier Med Vet International Supplier No. RXWATER1000INJV
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Sterile water for injection is suitable for use as a diluent for the preparation of pharmaceutical solutions. Single-dose vial contains no preservatives. Not suitable for IV injection without first having been made isotonic by addition of suitable solutes.