Learn More
Med Vet International Sterile Water For Injection, Diluent, 500mL, Each
Catalog No. 502828422
Encompass_Preferred
Sterile water for injection is suitable for use as a diluent for the preparation of pharmaceutical solutions. Single-dose vial contains no preservatives. Not suitable for IV injection without first having been made isotonic by addition of suitable solutes.
Catalog No. 50-282-8422 Supplier Med Vet International Supplier No. RXWATER500INJV
Sterile water for injection is suitable for use as a diluent for the preparation of pharmaceutical solutions. Single-dose vial contains no preservatives. Not suitable for IV injection without first having been made isotonic by addition of suitable solutes.