Learn More
AdipoGen Strobilurin B
SDP
Supplier:  AdipoGen BVT0312C500
Encompass_Preferred
Chemical. CAS 65105-52-4. Formula C17H19ClO4. MW 322.8. Isolated from Strobilurus sp. Antifungal. Respiration inhibitor. Blocks the electron transfer between cytochrome b and cytochrome c1.
Catalog No. 50-256-1993