Learn More
Cayman Chemical Strogen receptr B regIon 100uL
SDP
Catalog No. 502949405
Encompass_Preferred
A derivative of DL-homocysteine inhibits the growth of B. campestris L. sativa and E. utilis roots at 50 UM decreases the maximum rate of left ventricular developed pressure systolic left ventricular pressure and coronary flow in isolated rat hearts at 10 UM induces arteriosclerotic plaque formation in rabbits at 30 mg/kg for eight weeks has also been used as a precursor in the synthesis of thiolactone-containing monomers for use in polymer-based formaldehyde-scavenging coatings
Catalog No. 50-294-9405 Supplier Cayman Chemical Supplier No. 32768100 UL
A derivative of DL-homocysteine inhibits the growth of B. campestris L. sativa and E. utilis roots at 50 UM decreases the maximum rate of left ventricular developed pressure systolic left ventricular pressure and coronary flow in isolated rat hearts at 10 UM induces arteriosclerotic plaque formation in rabbits at 30 mg/kg for eight weeks has also been used as a precursor in the synthesis of thiolactone-containing monomers for use in polymer-based formaldehyde-scavenging coatings