Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strong Acid Solution, For Phosphorus, LabChem™

Catalog No. LC253701 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC253701 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC253701 Supplier LabChem, Inc. Supplier No. LC253701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Strong Acid Solution
Color Colorless
Name Note For Phosphorus
CAS Max % 58.43, 41.17, 0.4
CAS 7664-93-9, 7697-37-2
Linear Formula H2SO4
UN Number UN2796
Density 1.31g/mL
Solubility Information Soluble in water
Formula Weight 98.08
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.31g/mL
Viscosity 2.1cSt
Decomposition Information Sulfur compounds; Nitrogen oxides
Quantity 500 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.